3-(trans-4-Hydroxycyclohexylcarbamoyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(3-((trans-4-Hydroxycyclohexyl)carbamoyl)phenyl)boronic acid
BB-5072-1G
BB-5072-5G
COBBBB-5072-1GEA
497
EUR
COBBBB-5072-1G
COBBBB-5072-5G
3-(trans-4-Hydroxycyclohexylcarbamoyl)phenylboronsäure
3-(trans-4-Hydroxycyclohexylcarbamoyl)phenylboronsäure
Formel:
C₁₃H₁₈BNO₄ Molecular Weight: 263,0973 g/mol |
MDL:
MFCD09878335 CAS-Nummer: 957062-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...