4-(trans-4-Hydroxycyclohexylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-((trans-4-Hydroxycyclohexyl)carbamoyl)phenyl)boronic acid

BB-5062-5G BB-5062-1G
COBBBB-5062-5GEA 1700 EUR
COBBBB-5062-5G COBBBB-5062-1G
4-(trans-4-Hydroxycyclohexylcarbamoyl)phenylboronsäure
4-(trans-4-Hydroxycyclohexylcarbamoyl)phenylboronsäure
Formel: C₁₃H₁₈BNO₄
Molecular Weight: 263,0973 g/mol
MDL: MFCD09878333
CAS-Nummer: 957062-70-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR