3,5-Dichlor-2-hydroxybenzolsulfonsäure Natriumsalz
Lieferant: FLUOROCHEM
Synonyme:
Sodium 3,5-Dichloro-2-hydroxybenzenesulphonate, DHBS
001343-25G
001343-100G
FLUO001343-25GEA
0
EUR
FLUO001343-25G
FLUO001343-100G
3,5-Dichlor-2-hydroxybenzolsulfonsäure Natriumsalz
3,5-Dichlor-2-hydroxybenzolsulfonsäure Natriumsalz
Formel:
C₆H₃Cl₂NaO₄S Molecular Weight: 265.05 g/mol |
MDL:
MFCD00009798 CAS-Nummer: 54970-72-8 EINECS: 259-416-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...