3-Cyan-4-fluorbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

013064-5G 013064-25G
FLUO013064-5GEA 97.2 EUR
FLUO013064-5G FLUO013064-25G
3-Cyan-4-fluorbenzolsulfonylchlorid
3-Cyan-4-fluorbenzolsulfonylchlorid
Formel: C₇H₃ClFNO₂S
Molecular Weight: 219.62 g/mol
MDL: MFCD03093760
CAS-Nummer: 351003-23-1
UN: 3261
ADR: 8 I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR