2-Amino-5-(4-chlorphenyl)-4-thiazolcarbohydrazid

Lieferant: BIONET RESEARCH

1Y-0925.1 1Y-0925.10 1Y-0925.5 1Y-0925.500
BIOT1Y-0925.1EA 0 EUR
BIOT1Y-0925.1 BIOT1Y-0925.10 BIOT1Y-0925.5 BIOT1Y-0925.500
2-Amino-5-(4-chlorphenyl)-4-thiazolcarbohydrazid
2-Amino-5-(4-chlorphenyl)-4-thiazolcarbohydrazid
Formel: C₁₀H₉ClN₄OS
MDL: MFCD06200937
CAS-Nummer: 886361-54-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR