2-Amino-5-(2-chlorphenyl)-4-thiazolcarbonsäure
Lieferant: BIONET RESEARCH
12X-0929.1
12X-0929.10
12X-0929.5
12X-0929.500
BIOT12X-0929.1EA
0
EUR
BIOT12X-0929.1
BIOT12X-0929.10
BIOT12X-0929.5
BIOT12X-0929.500
2-Amino-5-(2-chlorphenyl)-4-thiazolcarbonsäure
2-Amino-5-(2-chlorophenyl)-4-thiazolecarboxylic acid
Formel:
C₁₀H₇ClN₂O₂S |
MDL:
MFCD06200928 CAS-Nummer: 886361-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...