2-Amino-5-(3-fluorphenyl)-4-thiazolcarbohydrazid

Lieferant: BIONET RESEARCH

1Y-0935.1 1Y-0935.10 1Y-0935.5 1Y-0935.500
BIOT1Y-0935.1EA 0 EUR
BIOT1Y-0935.1 BIOT1Y-0935.10 BIOT1Y-0935.5 BIOT1Y-0935.500
2-Amino-5-(3-fluorphenyl)-4-thiazolcarbohydrazid
2-Amino-5-(3-fluorphenyl)-4-thiazolcarbohydrazid
Formel: C₁₀H₉FN₄OS
MDL: MFCD06200940
CAS-Nummer: 886361-60-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR