3-(Cyanmethoxy)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)acetonitrile

PN-8675-5G PN-8675-1G
COBBPN-8675-5GEA 1410 EUR
COBBPN-8675-5G COBBPN-8675-1G
3-(Cyanmethoxy)phenylboronsäurepinakolester
3-(Cyanmethoxy)phenylboronsäurepinakolester
Formel: C₁₄H₁₈BNO₃
Molecular Weight: 259,1086 g/mol
MDL: MFCD06795674
CAS-Nummer: 936250-18-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR