4-(Cyanmethoxy)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)acetonitrile
PN-8676-5G
PN-8676-1G
COBBPN-8676-5GEA
451
EUR
COBBPN-8676-5G
COBBPN-8676-1G
4-(Cyanmethoxy)phenylboronsäurepinakolester
4-(Cyanmethoxy)phenylboronsäurepinakolester
Formel:
C₁₄H₁₈BNO₃ Molecular Weight: 259,1100 g/mol |
MDL:
MFCD06795675 CAS-Nummer: 475272-13-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...