3-(Cyanmethoxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-(Cyanomethoxy)phenyl)boronic acid

BB-9004-1G BB-9004-250M
COBBBB-9004-1GEA 685 EUR
COBBBB-9004-1G COBBBB-9004-250M
3-(Cyanmethoxy)phenylboronsäure
3-(Cyanmethoxy)phenylboronsäure
Formel: C₈H₈BNO₃
Molecular Weight: 176.9700 g/mol
MDL: MFCD09908169
CAS-Nummer: 947533-25-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR