2'-(Methylthio)biphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4849-5G CA-4849-1G
COBBCA-4849-5GEA 0 EUR
COBBCA-4849-5G COBBCA-4849-1G
2'-(Methylthio)biphenyl-3-carbonsäure
2'-(Methylthio)biphenyl-3-carbonsäure
Formel: C₁₄H₁₂O₂S
MDL: MFCD15143489
CAS-Nummer: 168618-43-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR