Isobornylacetat ≥94%

Lieferant: Thermo Fisher Scientific
Achtung

L11748.AE L11748.AP
L11748.AEEA 28.2 EUR
L11748.AE L11748.AP
Isobornylacetat ≥94%
Isobornylacetat

Purity: 94% CAS No.: 125-12-2 Molecular Formula: C12H20O2 Formula Weight: 196.29 Flash Point: 89°(192°F) Refractive Index: 1.4650 Boiling Point: 220-224° Beilstein Reference: 3197572 MDL No.: MFCD00135943

Formel: C₁₂H₂₀O₂
Molecular Weight: 196.29 g/mol
Siedepunkt: 220…224 °C
Dichte: 0.990
Flammpunkt: 89 °C (192 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00135943
CAS-Nummer: 125-12-2
EINECS: 204-727-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR