Tetrakis(trimethylsiloxy)silan

Lieferant: CHEMPUR SILANE

SILS15850.25 SILS15850.100
SILNSILS15850.25EA 0 EUR
SILNSILS15850.25 SILNSILS15850.100
Tetrakis(trimethylsiloxy)silan
Tetrakis(trimethylsiloxy)silan
Formel: C₁₂H₃₆O₄Si₅
MDL: MFCD00051587
CAS-Nummer: 3555-47-3
EINECS: 222-613-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR