Tetrakis(dimethylsiloxy)silan

Lieferant: CHEMPUR SILANE

Synonyme: Tetrakis(dimethylsilyl) orthosilicate

SILS15675.5 SILS15675.25 SILS15675.100
SILNSILS15675.5EA 0 EUR
SILNSILS15675.5 SILNSILS15675.25 SILNSILS15675.100
Tetrakis(dimethylsiloxy)silan
Tetrakis(dimethylsiloxy)silan
Formel: C₈H₂₈O₄Si₅
Molecular Weight: 328,73 g/mol
MDL: MFCD00053848
CAS-Nummer: 17082-47-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR