Tetrakis(trimethylsilyl)silan ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
TMSS
B21020.03
B21020.06
B21020.03EA
37.7
EUR
B21020.03
B21020.06
Tetrakis(trimethylsilyl)silan ≥98%
Tetrakis(trimethylsilyl)silan
Purity: 98%
CAS No.: 4098-98-0
Molecular Formula: Si[Si(CH3)3]4
Formula Weight: 320.85
Melting Point: 266-268° subl.
MDL No.: MFCD00054859
Formel:
C₁₂H₃₆Si₅ Molecular Weight: 320,842 g/mol Schmelzpunkt: 266…268 °C (Subl.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00054859 CAS-Nummer: 4098-98-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...