Tetrakis(trimethylsilyl)silan

Lieferant: CHEMPUR SILANE

SILS15900.1 SILS15900.5
SILNSILS15900.1EA 0 EUR
SILNSILS15900.1 SILNSILS15900.5
Tetrakis(trimethylsilyl)silan
Tetrakis(trimethylsilyl)silan
Formel: C₁₂H₃₆Si₅
MDL: MFCD00054859
CAS-Nummer: 4098-98-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR