(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP)
Lieferant: COMBI-BLOCKS
Synonyme:
1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tris(dimethylamino)phosphonium hexafluorophosphate(V), BOP reagent, Benzotriazol-1-yloxytris(dimethylamino)-phosphonium hexafluorophosphate
OS-7620-25G
OS-7620-100G
OS-7620-5G
COBBOS-7620-25GEA
0
EUR
COBBOS-7620-25G
COBBOS-7620-100G
COBBOS-7620-5G
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP)
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP)
Formel:
C₁₂H₂₂F₆N₆OP₂ Molecular Weight: 442,28 g/mol |
MDL:
MFCD00011948 CAS-Nummer: 56602-33-6 EINECS: 260-279-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...