5-Amino-3-brom-2-methoxypyridin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-6-methoxypyridin-3-amine

PY-7865-25G PY-7865-5G
COBBPY-7865-25GEA 204 EUR
COBBPY-7865-25G COBBPY-7865-5G
5-Amino-3-brom-2-methoxypyridin
5-Amino-3-brom-2-methoxypyridin
Formel: C₆H₇BrN₂O
Molecular Weight: 203,0400 g/mol
MDL: MFCD06659518
CAS-Nummer: 53242-18-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR