2-Amino-6-methoxy-3-nitropyridin

Lieferant: COMBI-BLOCKS

Synonyme: 6-Methoxy-3-nitropyridin-2-amine

OS-7409-5G OS-7409-25G OS-7409-100G
COBBOS-7409-5GEA 26.2 EUR
COBBOS-7409-5G COBBOS-7409-25G COBBOS-7409-100G
2-Amino-6-methoxy-3-nitropyridin
2-Amino-6-methoxy-3-nitropyridine
Formel: C₆H₇N₃O₃
Molecular Weight: 169,14 g/mol
MDL: MFCD03206481
CAS-Nummer: 73896-36-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR