(1-Ethoxycarbonylethyliden)triphenylphosphoran ca. 94% balance TPPO
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(Carbethoxyethylidene)triphenylphosphorane, Ethyl 2-(triphenylphosphoranylidene)propanoate
146080050
146080250
ACRO146080050EA
36
EUR
ACRO146080050
ACRO146080250
(1-Ethoxycarbonylethyliden)triphenylphosphoran ca. 94% balance TPPO
(1-Ethoxycarbonylethyliden)triphenylphosphoran
Formel:
C₂₃H₂₃O₂P Molecular Weight: 362,41 g/mol Schmelzpunkt: 157…159 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00009160 CAS-Nummer: 5717-37-3 EINECS: 227-215-4 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Yellow Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ca. 94 % |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...