(1-Ethoxycarbonylethyliden)triphenylphosphoran ≥97%
Lieferant: Thermo Scientific
Synonyme:
(Carbethoxyethylidene)triphenylphosphorane, Ethyl 2-(triphenylphosphoranylidene)propanoate
A15619.09
A15619.14
A15619.18
A15619.30
A15619.09EA
66.7
EUR
A15619.09
A15619.14
A15619.18
A15619.30
(1-Ethoxycarbonylethyliden)triphenylphosphoran ≥97%
(1-Ethoxycarbonylethyliden)triphenylphosphoran
Purity: 97%
CAS No.: 5717-37-3
Molecular Formula: C23H23O2P
Formula Weight: 362.41
Storage Temperature: 2°C to 8°C
Melting Point: 154-160°
Beilstein Reference: 757112
MDL No.: MFCD00009160
Formel:
C₂₃H₂₃O₂P Molecular Weight: 362,41 g/mol Schmelzpunkt: 154…160 °C |
MDL:
MFCD00009160 CAS-Nummer: 5717-37-3 EINECS: 227-215-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...