Lanthantriacetat Sequihydrat ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden)

Lieferant: Thermo Scientific

Synonyme: Lanthanum(III) acetate sesquihydrate

11263.18 11263.30
11263.18EA 117 EUR
11263.18 11263.30
Lanthantriacetat Sequihydrat ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lanthantriacetat Sequihydrat

Purity: 99.99% CAS No.: 25721-92-0 Molecular Formula: C6H9LaO6.1.5H2O Formula Weight: 343.05 (316.03anhy) Physical Form: Crystalline Sensitivity: Hygroscopic MDL No.: MFCD00013037

Formel: C₆H₉LaO₆·1.5H₂O
Molecular Weight: 343,05 g/mol (316,03 anhy)
Schmelzpunkt: <200 °C (-1,5H₂O)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013037
CAS-Nummer: 25721-92-0
EINECS: 213-034-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR