(2,6-Dichlorbenzyl)tripropylphosphoniumchlorid

Lieferant: Thermo Scientific

CD03615.250 CD03615.1
MAYBCD03615.250EA 0 EUR
MAYBCD03615.250 MAYBCD03615.1
(2,6-Dichlorbenzyl)tripropylphosphoniumchlorid
(2,6-Dichlorbenzyl)tripropylphosphoniumchlorid
Formel: C₁₆H₂₆Cl₃P
Molecular Weight: 355,71 g/mol
CAS-Nummer: 646497-56-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR