(2,3,4,5,6-Pentamethylbenzyl)tripropylphosphoniumchlorid

Lieferant: Thermo Scientific

Synonyme: (2,3,4,5,6-Pentamethylbenzyl)(tripropyl)phosphonium chloride

CD03614.250 CD03614.1
MAYBCD03614.250EA 0 EUR
MAYBCD03614.250 MAYBCD03614.1
(2,3,4,5,6-Pentamethylbenzyl)tripropylphosphoniumchlorid
(2,3,4,5,6-Pentamethylbenzyl)tripropylphosphoniumchlorid
Formel: C₂₁H₃₈ClP
Molecular Weight: 356,95 g/mol
MDL: MFCD02682093
CAS-Nummer: 646497-54-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR