Benzyl(tributyl)phosphoniumchlorid

Lieferant: Thermo Scientific

JFD04037.250 JFD04037.1
MAYBJFD04037.250EA 0 EUR
MAYBJFD04037.250 MAYBJFD04037.1
Benzyl(tributyl)phosphoniumchlorid
Benzyl(tributyl)phosphoniumchlorid
Formel: C₁₉H₃₄ClP
CAS-Nummer: 1224-56-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR