Methyl-4'-aminobiphenyl-3-carboxylat

Lieferant: COMBI-BLOCKS

OT-8011-5G OT-8011-1G
COBBOT-8011-5GEA 1090 EUR
COBBOT-8011-5G COBBOT-8011-1G
Methyl-4'-aminobiphenyl-3-carboxylat
Methyl-4'-aminobiphenyl-3-carboxylat
Formel: C₁₄H₁₃NO₂
MDL: MFCD04039111
CAS-Nummer: 400747-22-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR