2-Chlor-4,5-difluorbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

CFF907. 005519-1G 005519-5G
CHMPCFF907.EA 0 EUR
CHMPCFF907. FLUO005519-1G FLUO005519-5G
2-Chlor-4,5-difluorbenzolsulfonylchlorid
2-Chlor-4,5-difluorbenzolsulfonylchlorid
Formel: C₆H₂Cl₂F₂O₂S
MDL: MFCD02091388
CAS-Nummer: 67475-58-5
UN: 1760
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR