4-Fluor-3-methoxybenzolsulfonylchlorid 95%

Lieferant: Apollo Scientific
Gefahr

PC10594-1G
APOSPC10594-1GEA 101 EUR
APOSPC10594-1G
4-Fluor-3-methoxybenzolsulfonylchlorid 95%
4-Fluor-3-methoxybenzolsulfonylchlorid
Formel: C₇H₆ClFO₃S
MDL: MFCD07368708
CAS-Nummer: 887266-97-9
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR