6-Chlor-4-nitro-o-kresol
Lieferant: Thermo Scientific
Synonyme:
2-Chloro-6-methyl-4-nitrophenol
BTB05378.250
BTB05378.1
MAYBBTB05378.250EA
0
EUR
MAYBBTB05378.250
MAYBBTB05378.1
6-Chlor-4-nitro-o-kresol
6-Chlor-4-nitro-o-kresol
Formel:
C₇H₆ClNO₃ |
MDL:
MFCD00047740 CAS-Nummer: 4102-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...