4-Chlor-6-nitro-m-kresol

Lieferant: CHEMPUR

FL13478.POR 013478-5G
CHMPFL13478.POREA 0 EUR
CHMPFL13478.POR FLUO013478-5G
4-Chlor-6-nitro-m-kresol
4-Chlor-6-nitro-m-kresol
Formel: C₇H₆ClNO₃
MDL: MFCD00007114
CAS-Nummer: 7147-89-9
EINECS: 230-461-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR