trans-2-Hexenylacetat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ300

W256404-100G-K W256404-1KG-K W256404-5KG-K W256404-9KG-K
SIALW256404-100G-KEA 85.8 EUR
SIALW256404-100G-K SIALW256404-1KG-K SIALW256404-5KG-K SIALW256404-9KG-K
trans-2-Hexenylacetat, Sigma-Aldrich®
trans-2-Hexenylacetat
Formel: C₈H₁₄O₂
Molecular Weight: 142.20 g/mol
MDL: MFCD00009474
CAS-Nummer: 2497-18-9
EINECS: 219-680-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR