(2-Chlorbenzyl)triphenylphosphoniumchlorid
Lieferant: Thermo Scientific
JFD04061.250
JFD04061.1
MAYBJFD04061.250EA
0
EUR
MAYBJFD04061.250
MAYBJFD04061.1
(2-Chlorbenzyl)triphenylphosphoniumchlorid
(2-Chlorbenzyl)triphenylphosphoniumchlorid
Formel:
C₂₅H₂₁Cl₂P Molecular Weight: 423,3140 g/mol |
MDL:
MFCD00054629 CAS-Nummer: 18583-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...