4-Amino-2-methoxypyridin

Lieferant: COMBI-BLOCKS

PY-7373-25G PY-7373-5G
COBBPY-7373-25GEA 0 EUR
COBBPY-7373-25G COBBPY-7373-5G
4-Amino-2-methoxypyridin
4-Amino-2-methoxypyridine
Formel: C₆H₈N₂O
MDL: MFCD06738657
CAS-Nummer: 20265-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR