3-Chlor-2,6-difluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Chloro-2,6-difluorophenyl)boronic acid

BB-8450-1G BB-8450-5G
COBBBB-8450-1GEA 170 EUR
COBBBB-8450-1G COBBBB-8450-5G
3-Chlor-2,6-difluorphenylboronsäure
3-Chlor-2,6-difluorphenylboronsäure
Formel: C₆H₄BClF₂O₂
Molecular Weight: 192.3556 g/mol
MDL: MFCD04038753
CAS-Nummer: 1031226-45-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR