3-Chlor-5-fluorphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Chloro-5-fluorobenzeneboronic acid, 3-Chloro-5-fluorophenlyboronic acid
BB-2390-5G
BB-2390-1G
COBBBB-2390-5GEA
127
EUR
COBBBB-2390-5G
COBBBB-2390-1G
3-Chlor-5-fluorphenylboronsäure
3-Chlor-5-fluorphenylboronsäure
Formel:
C₆H₅BClFO₂ Molecular Weight: 174,37 g/mol |
MDL:
MFCD06801741 CAS-Nummer: 328956-61-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...