4-Chlor-2,6-difluorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8389-5G BB-8389-1G
COBBBB-8389-5GEA 1050 EUR
COBBBB-8389-5G COBBBB-8389-1G
4-Chlor-2,6-difluorphenylboronsäure
4-Chlor-2,6-difluorphenylboronsäure
Formel: C₆H₄BClF₂O₂
Molecular Weight: 192,3600 g/mol
MDL: MFCD03095138
CAS-Nummer: 925911-61-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR