2-Amino-1-naphthalinsulfonsäure 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Aminonaphthalene-1-sulphonic acid
A18644.36
A18644.0E
A18644.36EA
54.8
EUR
A18644.36
A18644.0E
2-Amino-1-naphthalinsulfonsäure 98%
2-Amino-1-naphthalenesulphonic acid
Purity: 98%
CAS No.: 81-16-3
Molecular Formula: C10H9NO3S
Formula Weight: 223.25
Beilstein Reference: 613084
Merck Reference: 14,6406
MDL No.: MFCD00003988
Formel:
C₁₀H₉NO₃S Molecular Weight: 223.25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003988 CAS-Nummer: 81-16-3 EINECS: 201-331-5 Merck Index: 14,6406 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...