2,6-Dichlor-4-(trifluormethyl)benzolsulfonylchlorid
Lieferant: FLUOROCHEM
004743-25G
004743-5G
FLUO004743-25GEA
306
EUR
FLUO004743-25G
FLUO004743-5G
2,6-Dichlor-4-(trifluormethyl)benzolsulfonylchlorid
2,6-Dichlor-4-(trifluormethyl)benzolsulfonylchlorid
Formel:
C₇H₂Cl₃F₃O₂S |
MDL:
MFCD00220929 CAS-Nummer: 175205-76-2 UN: 3261 ADR: 8,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...