4-Nitro-3-(trifluormethyl)benzolsulfonylchlorid
Lieferant: CHEMPUR
Synonyme:
4-Nitro-3-(trifluoromethyl)benzene-1-sulphonyl chloride
FL13088.POR
013088-1G
013088-5G
CHMPFL13088.POREA
0
EUR
CHMPFL13088.POR
FLUO013088-1G
FLUO013088-5G
4-Nitro-3-(trifluormethyl)benzolsulfonylchlorid
4-Nitro-3-(trifluormethyl)benzolsulfonylchlorid
Formel:
C₇H₃ClF₃NO₄S Molecular Weight: 289,62 g/mol |
MDL:
MFCD03094387 CAS-Nummer: 39234-83-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...