1,2-Dichlor-4-(methylsulfonyl)benzol

Lieferant: COMBI-BLOCKS

OT-0636-25G OT-0636-1G OT-0636-5G
COBBOT-0636-25GEA 335 EUR
COBBOT-0636-25G COBBOT-0636-1G COBBOT-0636-5G
1,2-Dichlor-4-(methylsulfonyl)benzol
1,2-Dichloro-4-methylsulphonylbenzene
Formel: C₇H₆Cl₂O₂S
MDL: MFCD00025063
CAS-Nummer: 38452-47-0
EINECS: 253-942-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR