4-Brom-2,5-dimethoxybenzaldehyd ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

B25500.14 B25500.06
B25500.14EA 230 EUR
B25500.14 B25500.06
4-Brom-2,5-dimethoxybenzaldehyd ≥98%
4-Bromo-2,5-dimethoxybenzaldehyde

Purity: 98+% CAS No.: 31558-41-5 Molecular Formula: C9H9BrO3 Formula Weight: 245.08 Melting Point: 132-135° MDL No.: MFCD00156964

Formel: C₉H₉BrO₃
Molecular Weight: 245.08 g/mol
Schmelzpunkt: 132…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00156964
CAS-Nummer: 31558-41-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR