1-Brom-4,5-dichlor-2-nitrobenzol

Lieferant: COMBI-BLOCKS

AN-1208-1G AN-1208-5G
COBBAN-1208-1GEA 101 EUR
COBBAN-1208-1G COBBAN-1208-5G
1-Brom-4,5-dichlor-2-nitrobenzol
1-Brom-4,5-dichlor-2-nitrobenzol
Formel: C₆H₂BrCl₂NO₂
Molecular Weight: 270,9000 g/mol
MDL: MFCD07780670
CAS-Nummer: 93361-94-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR