1-Brom-3-chlor-5-nitrobenzol

Lieferant: COMBI-BLOCKS

AN-1198-5G AN-1198-1G
COBBAN-1198-5GEA 130 EUR
COBBAN-1198-5G COBBAN-1198-1G
1-Brom-3-chlor-5-nitrobenzol
1-Bromo-3-chloro-5-nitrobenzene
Formel: C₆H₃BrClNO₂
Molecular Weight: 236,4500 g/mol
MDL: MFCD08275708
CAS-Nummer: 219817-43-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR