3-Amino-2-(4-methoxyphenyl)pyridin

Lieferant: BIONET RESEARCH

5Y-0701.5 5Y-0701.1 5Y-0701.10 5Y-0701.500
BIOT5Y-0701.5EA 0 EUR
BIOT5Y-0701.5 BIOT5Y-0701.1 BIOT5Y-0701.10 BIOT5Y-0701.500
3-Amino-2-(4-methoxyphenyl)pyridin
3-Amino-2-(4-methoxyphenyl)pyridine
Formel: C₁₂H₁₂N₂O
MDL: MFCD06411622
CAS-Nummer: 663918-44-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR