5-Amino-2-(4-methoxyphenyl)pyridin

Lieferant: BIONET RESEARCH

Synonyme: 6-(4-Methoxyphenyl)pyridin-3-amine

4Y-0705.500 4Y-0705.10 4Y-0705.1 4Y-0705.5
BIOT4Y-0705.500EA 0 EUR
BIOT4Y-0705.500 BIOT4Y-0705.10 BIOT4Y-0705.1 BIOT4Y-0705.5
5-Amino-2-(4-methoxyphenyl)pyridin
5-Amino-2-(4-methoxyphenyl)pyridin
Formel: C₁₂H₁₂N₂O
MDL: MFCD05864840
CAS-Nummer: 52057-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR