4-Hydroxy-2,6-dimethoxybenzaldehyd

Lieferant: COMBI-BLOCKS

Synonyme: 2,6-Dimethoxy-4-hydroxybenzaldehyde

BD-0035-5G BD-0035-1G
COBBBD-0035-5GEA 181 EUR
COBBBD-0035-5G COBBBD-0035-1G
4-Hydroxy-2,6-dimethoxybenzaldehyd
4-Hydroxy-2,6-dimethoxybenzaldehyd
Formel: C₉H₁₀O₄
Molecular Weight: 182.1733 g/mol
MDL: MFCD01407645
CAS-Nummer: 22080-96-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR