1-Chlor-4,5-difluor-2-nitrobenzol

Lieferant: CHEMPUR

Synonyme: 2-Chloro-4,5-difluoronitrobenzene

FL13796.POR 013796-1G 013796-5G
CHMPFL13796.POREA 0 EUR
CHMPFL13796.POR FLUO013796-1G FLUO013796-5G
1-Chlor-4,5-difluor-2-nitrobenzol
1-Chlor-4,5-difluor-2-nitrobenzol
Formel: C₆H₂ClF₂NO₂
MDL: MFCD04973764
CAS-Nummer: 771-76-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR