1,2,4,5-Tetrakis(brommethyl)benzol 95%
Lieferant: Thermo Scientific
|
Gefahr
|
370920050
370920250
ACRO370920050EA
114
EUR
ACRO370920050
ACRO370920250
1,2,4,5-Tetrakis(brommethyl)benzol 95%
1,2,4,5-Tetrakis(brommethyl)benzol
Formel:
C₁₀H₁₀Br₄ Molecular Weight: 449,8 g/mol Schmelzpunkt: 156…162 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009664 CAS-Nummer: 15442-91-8 UN: 1759 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | Off-white to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 155 to 161 °C |
HPLC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...