2-Acetamidoethanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Acetylethanolamine

M-Clarity™ Qualitätsstufe = MQ100

100455-25G 100455-500G
SIAL100455-25GEA 50 EUR
SIAL100455-25G SIAL100455-500G
2-Acetamidoethanol, Sigma-Aldrich®
2-Acetamidoethanol
Formel: C₄H₉NO₂
Molecular Weight: 103,12 g/mol
MDL: MFCD00002836
CAS-Nummer: 142-26-7
EINECS: 205-530-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR