1-(Ammoniooxy)propanchlorid
Lieferant: BIONET RESEARCH
12H-939.500
12H-939.1
12H-939.5
12H-939.10
BIOT12H-939.500EA
0
EUR
BIOT12H-939.500
BIOT12H-939.1
BIOT12H-939.5
BIOT12H-939.10
1-(Ammoniooxy)propanchlorid
1-(Ammoniooxy)propanchlorid
Formel:
C₃H₁₀ClNO |
MDL:
MFCD00231456 CAS-Nummer: 6084-54-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...